Learn More
Strem, An Ascensus Company CAS# 1303-86-2. 1kg. Boron oxide (99.6%-B). MFCD00011315
SDP
Catalog No. 5090113723
Encompass
CAS# 1303-86-2. 1kg. Boron oxide (99.6%-B). MFCD00011315. Molecular Weight: 69.62. Molecular Formula: B2O3. Color/form: white pwdr. Strem# 93-0526. http://www.strem.com/catalog/v/93-0526/
Catalog No. 50-901-13723 Supplier Strem, An Ascensus Company Supplier No. 9305261000G
CAS# 1303-86-2. 1kg. Boron oxide (99.6%-B). MFCD00011315. Molecular Weight: 69.62. Molecular Formula: B2O3. Color/form: white pwdr. Strem# 93-0526. http://www.strem.com/catalog/v/93-0526/