Learn More
Strem, An Ascensus Company CAS# 1303-86-2. 50g. Boron oxide (99.9+%-B). MFCD00011315
SDP
Catalog No. 5090113724
Encompass
CAS# 1303-86-2. 50g. Boron oxide (99.9+%-B). MFCD00011315. Molecular Weight: 69.62. Molecular Formula: B2O3. Color/form: white pwdr. Strem# 93-0527. http://www.strem.com/catalog/v/93-0527/
Catalog No. 50-901-13724 Supplier Strem, An Ascensus Company Supplier No. 93052750G
CAS# 1303-86-2. 50g. Boron oxide (99.9+%-B). MFCD00011315. Molecular Weight: 69.62. Molecular Formula: B2O3. Color/form: white pwdr. Strem# 93-0527. http://www.strem.com/catalog/v/93-0527/