Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron tribromide, 1M solution in methylene chloride, AcroSeal™

Catalog No. p-4440132
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427101000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427101000 Supplier Thermo Scientific Chemicals Supplier No. 427101000
Only null left
Add to Cart

CAS: 10294-33-4 | BBr3 | 250.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron tribromide
CAS 10294-33-4, 75-09-2
Quantity 100 mL
Molecular Formula BBr3
Linear Formula BBr3
MDL Number MFCD00011312
Merck Index 15,1349
Synonym boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in alcohol and ccl4
InChI Key ILAHWRKJUDSMFH-UHFFFAOYSA-N
SMILES BrB(Br)Br
IUPAC Name tribromoborane
Molecular Weight (g/mol) 250.52
PubChem CID 25134
Formula Weight 250.52
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.467
Density 1.4670g/mL
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.