Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Tribromide Dimethyl Sulfide Complex Solution

Catalog No. 247383100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-738-3100ML 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 24-738-3100ML Supplier Sigma Aldrich Supplier No. 247383100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Precursor of HIV protease inhibitor KNI 764 using cation-exchange resin mediated Mannich reactionReagent for:Baylis-Hillman reactions

Specifications

CAS 100-58-3
Density 1.134 g/mL at 25°C
Molecular Formula C6H5BrMg
Linear Formula C6H5MgBr
MDL Number MFCD00000038
Quantity 200 mL
Synonym Dimethyl sulfide-tribromoborane; Tribromoborane-methyl sulfide
Molecular Weight (g/mol) 181.31 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.