Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Tribromide Solution

Catalog No. 211230100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-123-0100ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 21-123-0100ML Supplier Sigma Aldrich Supplier No. 211230100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Drug intermediate 6-nitro-L-DOPALuminescent polystyrene derivatives with sterically protected carbazolylborane moietiesHigh-quality boron-doped graphene via Wurtz-type reductive coupling reactionMercapto-(+)-methamphetamine haptens for synthesis of (+)-methamphetamine conjugate vaccines with improved epitope densitiesMicrometer-sized organic molecule-DNA hybrid structuresBorane complexes via electrophilic aromatic borylation reactionsA 5-HT2C receptor agonistBiphenyl-derivatives possessing tertiary amino groups as +â-secretase(BACE1) inhibitors for the treatment of Alzheimer'sas diseaseA highly near-IR region fluorescent p-extended boron aza-dipyrromethene moiety unitTetrahydroisoquinoline derivatives via intramolecular cyclization of methoxy-substituted N-phenethylimides

Specifications

CAS 3699-66-9
Boiling Point 143°C to 144°C (12 mmHg, lit.)
MDL Number MFCD02683470
Quantity 5 g
Synonym Tribromoboron
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.