Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Tribromide Solution

Catalog No. 403857100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-385-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 40-385-7100ML Supplier Sigma Aldrich Supplier No. 403857100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Drug intermediate 6-nitro-L-DOPALuminescent polystyrene derivatives with sterically protected carbazolylborane moietiesHigh-quality boron-doped graphene via Wurtz-type reductive coupling reactionMercapto-(+)-methamphetamine haptens for synthesis of (+)-methamphetamine conjugate vaccines with improved epitope densitiesMicrometer-sized organic molecule-DNA hybrid structuresBorane complexes via electrophilic aromatic borylation reactionsA 5-HT2C receptor agonistBiphenyl-derivatives possessing tertiary amino groups as +â-secretase(BACE1) inhibitors for the treatment of Alzheimer'sas diseaseA highly near-IR region fluorescent p-extended boron aza-dipyrromethene moiety unitTetrahydroisoquinoline derivatives via intramolecular cyclization of methoxy-substituted N-phenethylimides

Specifications

CAS 929-75-9
Molecular Formula C8H20N2O3
Linear Formula C8H20N2O3
MDL Number MFCD00628164
Quantity 100 mL
Synonym Tribromoboron
Molecular Weight (g/mol) 192.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.