Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron Trichloride (ca. 13% in p-Xylene, ca. 1.0mol/L), TCI America™
SDP

Catalog No. B3968100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3968100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3968100ML Supplier TCI America Supplier No. B3968100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10294-34-5
Molecular Formula BCl3
Molecular Weight (g/mol) 117.16
MDL Number MFCD00011313
InChI Key FAQYAMRNWDIXMY-UHFFFAOYSA-N
Synonym boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
PubChem CID 25135
IUPAC Name trichloroborane
SMILES ClB(Cl)Cl

Specifications

Color Yellow
Quantity 100 mL
UN Number 2920
Formula Weight 117.16
Physical Form Liquid
Chemical Name or Material Boron Trichloride (ca. 13% in p-Xylene, ca. 1.0mol/L)
EINECSNumber (1)-0042
RTECSNumber ED1925000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.