Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Trichloride Solution

Catalog No. 1789342L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
2 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-893-42L 2 L
17-893-4800ML 800 mL
17-893-4100ML 100 mL
17-893-425ML 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 17-893-42L Supplier Sigma Aldrich Supplier No. 1789342L
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron trichloride is a boron halide. It participates in the hydroboration of alkenes. It forms strongly polarized donor-acceptor complexes with trimethyl- and triethylsilyl triflate. Boron trichloride assisted reaction between acid chlorides and alkynyltrifluoroborate salts have been reported to form ynones.For preparing methyl esters of fatty acids and for transesterification of triglycerides.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 10294-34-5
Density 1.326 g/mL at 25°C
Molecular Formula BCl3
Linear Formula BCl3
MDL Number MFCD00011313
Quantity 2 L
Molecular Weight (g/mol) 117.17 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.