Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Trichloride Solution

Catalog No. 678732800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-873-2800ML 800 mL
67-873-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 67-873-2800ML Supplier Sigma Aldrich Supplier No. 678732800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron trichloride (BCl3) is a boron halide typically used as a reagent in organic synthesis for the cleavage of C-O bonds in ethers.Boron trichloride (BCl3) can be used as a reactant in the preperation of borazine-linked polymer [BLP-10(Cl)] by thermal decomposition with benzidine for gas storage and purification.

Specifications

CAS 10294-34-5
Density 0.909 g/mL at 25°C
Molecular Formula BCl3
Linear Formula BCl3
MDL Number MFCD00011313
Quantity 800 mL
Molecular Weight (g/mol) 117.17 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.