Learn More
Supelco Inc Boron trifluoride-methanol solution | 373-57-9 | MFCD00071635 | 500mL
Catalog No. NC0599135
Encompass
Boron trifluoride-methanol solution | 14% in methanol | MW: 99.85 | 373-57-9 | MFCD00071635 | 500mL
Catalog No. NC0599135 Supplier Supelco Inc Supplier No. B1252500ML
Boron trifluoride-methanol solution | 14% in methanol | MW: 99.85 | 373-57-9 | MFCD00071635 | 500mL