Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trifluoride, 99+%, Thermo Scientific Chemicals

Catalog No. p-7049791
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2287422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA2287422 Supplier Thermo Scientific Chemicals Supplier No. 02287422
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7637-07-2
Molecular Formula BF3
Molecular Weight (g/mol) 67.81
MDL Number MFCD00011316
InChI Key WTEOIRVLGSZEPR-UHFFFAOYSA-N
Synonym boron trifluoride, borane, trifluoro, boron fluoride, trifluoroboron, fluorure de bore, fluorure de bore french, unii-7jgd48px8p, 7jgd48px8p, trifluoro boron
PubChem CID 6356
ChEBI CHEBI:33093
IUPAC Name trifluoroborane
SMILES B(F)(F)F

Specifications

Boiling Point -100°C
Melting Point -127°C
Color Colorless
Physical Form Gas
Quantity 100 g
UN Number UN1008
Merck Index 14,1349
Formula Weight 67.81
Percent Purity ≥99%
Odor Pungent
Sensitivity Moisture sensitive
Density 3.076 g/mL
Chemical Name or Material Boron trifluoride
Show More Show Less
Hazard Category H280-H314-H330-H373
Hazard Statement GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 2
EINECSNumber 237-383-0
RTECSNumber ED2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.