Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Trifluoride Acetic Acid Complex

Catalog No. 359955250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-995-5250ML 250 mL
35-995-51L 1 L
35-995-55ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 35-995-5250ML Supplier Sigma Aldrich Supplier No. 359955250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron Trifluoride Acetic Acid Complex

Specifications

CAS 373-61-5
Density 1.353 g/mL at 25°C
Molecular Formula C4H8BF3O4
Linear Formula BF3 -+ 2CH3COOH
Refractive Index n20/D 1.368
Quantity 250 mL
MDL Number MFCD00036359
Molecular Weight (g/mol) 187.91 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.