Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trifluoride diethyl etherate, 46.5% BF{3} min, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA4179636
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4179636 500 g
AA4179622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4179636 Supplier Thermo Scientific Chemicals Supplier No. 04179636
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol

Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
PubChem CID 8000
SMILES FB(F)F.CCOCC

Specifications

Boiling Point 125°C to 126°C
Melting Point -60°C
Physical Form Liquid
Quantity 500 g
Linear Formula BF3·O(CH2CH3)2
UN Number UN2604
Beilstein 3909607
Flash Point 47°C (116°F)
Merck Index 14,1350
Solubility Information Miscible with ether and alcohol.
IUPAC Name ethoxyethane; trifluoroborane
Formula Weight 141.93
Concentration or Composition (by Analyte or Components) 46.5% BF3 min.
Odor Acrid
Packaging ChemSeal™ bottles
Refractive Index 1.344
Sensitivity Moisture sensitive
Density 1.130 g/mL
Chemical Name or Material Boron trifluoride diethyl etherate
Show More Show Less
Hazard Category H226-H314-H332-H372
Hazard Statement GHS H Statement
H331-H372-H314-H318-H226-H302
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIFLUORIDE DIETHYL ETHERATE
EINECSNumber 203-689-8
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.