Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trifluoride dimethyl etherate

Catalog No. AC204060500
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC204060500 50 mL Glass bottle
AC204062500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC204060500 Supplier Thermo Scientific Chemicals Supplier No. 204060500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 353-42-4 | C2H6BF3O | 113.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 353-42-4
Molecular Formula C2H6BF3O
Molecular Weight (g/mol) 113.87
MDL Number MFCD00013192
InChI Key KTPWETRNSUKEME-UHFFFAOYSA-N
Synonym boron trifluoride dimethyl etherate, boron trifluoride methyl etherate, dimethyl ether trifluoroboron, dimethyloxonio trifluoro boranuide, boron trifluoride dimethyl ether complex, dimethyloxo trifluoro-??-borane
PubChem CID 2724165
IUPAC Name dimethyloxonio(trifluoro)boranuide
SMILES COC.FB(F)F

Specifications

Melting Point -14.0°C
Color Brown
Density 1.2300g/mL
Boiling Point 126.0°C to 127.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3070 to 1.3090
Linear Formula (CH3)2O·BF3
Quantity 50 mL
Specific Gravity 1.23
Solubility Information Solubility in water: reacts
Formula Weight 113.87
Percent Purity ∼60% (BF3)
Physical Form Liquid, Fuming In Moist Air
Chemical Name or Material Boron trifluoride dimethyl etherate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-532-1
RTECSNumber ED8400000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.