Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Trifluoride Methanol Complex Solution

Catalog No. 61626500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-626-500ML 500 mL
61-626-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 61-626-500ML Supplier Sigma Aldrich Supplier No. 61626500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron trifluoride-methanol complex solution can be used for the deprotection of acetylated amines of cyanine dyes to form products with free amino groups. It may be used for the conversion of sulfur-containing phosphoramidates to the corresponding phosphate methyl ester.

Specifications

CAS 2802-68-8
Density 0.859 g/mL at 25°C
Molecular Formula C2H8BF3O2
Linear Formula BF3 -+ 2CH3OH
MDL Number MFCD00071635
Quantity 500 mL
Molecular Weight (g/mol) 131.89 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.