Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trifluoride-Methanol Solution, ∼10 (∼1.3 M), MilliporeSigma™ Supelco™

Catalog No. 111020687 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
100 mL
10 x 1 mL
250 mL
1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0687 1 mL
11-102-0140 10 x 1 mL
11-102-0608 10 mL
11-102-0334 100 mL
11-101-9291 250 mL
11-101-8152 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 11-102-0687 Supplier MilliporeSigma Supelco Supplier No. 157161ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00071635

Specifications

CAS 373-57-9
Quantity 1 mL
Linear Formula BF3 · MeOH
Flash Point 9°C
Formula Weight 99.85
Grade GC Derivatization
Concentration or Composition (by Analyte or Components) 10% (1.3 M)
Density 0.86 g/mL (at 20°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.