Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Trifluoride-Methanol Solution

Catalog No. 13482150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
250 mL
4 x 25 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-482-150ML 50 mL
13-482-1250ML 250 mL
13-482-125ML 4 x 25 mL
13-482-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 13-482-150ML Supplier Sigma Aldrich Supplier No. 13482150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the GC analysis of ritalinic acid. Alternative to Cl2CHCO2H for synthesis of purine rich oligomers.Lewis acid used in the preparation of geminal bis-peroxides from ketals and enol ethers with t-butyl hydroperoxide. Reagent used for the mild deacetylation of amines.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 373-57-9
Density 1.203 g/mL at 25°C
Boiling Point 59°C (4 mmHg)
Molecular Formula CH4BF3O
Linear Formula BF3 -+ MeOH
MDL Number MFCD00071635
Quantity 50 mL
Molecular Weight (g/mol) 99.85 g/mol
Biological Activity Eyes, Kidney, Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.