Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron trifluoride-methanol solution

Catalog No. B1252100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1252100ML 100 mL
B1252250ML 250 mL
B12525ML 5 mL
B12521L 1 L
B1252500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. B1252100ML Supplier Sigma Aldrich Supplier No. B1252100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron trifluoride-methanol (BF-M) is an esterification reagent. BF-M is a powerful acidic catalyst for the esterification of the fatty acids. It easily cleaves acetylated dyes resulting in free amino groups. BF-M catalyzed trans esterification of vegetable oils forms biodiesel.Used in the deactylation of acetanilides.It is also used to study the synthesis of amino group containing cyanine dyes of different types and to prepare fatty acid methyl esters.

Specifications

CAS 373-57-9
Density 0.859 g/mL (at 25°C)
Molecular Formula CH4BF3O
Linear Formula BF3 -+ MeOH
Quantity 100 mL
MDL Number MFCD00071635
Synonym Trifluoroborane
Molecular Weight (g/mol) 99.85
Percent Purity 12-16% (NaOH, titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.