Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Trifluoride Methyl Sulfide Complex

Catalog No. 261998100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-199-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 26-199-8100ML Supplier Sigma Aldrich Supplier No. 261998100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron trifluoride methyl sulfide complex is a common reagent used for dealkylation of ethers. It can also be used to synthesize boron trifluoride complex with phosphine ligands.

Specifications

CAS 18162-84-0
Density 0.873 g/mL at 25°C
Boiling Point 222°C to 225°C
Molecular Formula C10H23ClSi
Linear Formula CH3(CH2)7Si(CH3)2Cl
Quantity 100 mL
MDL Number MFCD00000511
Synonym Dimethyl sulfide-trifluoroborane; Trifluoroborane-methyl sulfide
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.