Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Trifluoride Tetrahydrofuran Complex

Catalog No. 434280100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-428-0100ML 100 mL
43-428-0500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-428-0100ML Supplier Sigma Aldrich Supplier No. 434280100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron Trifluoride Tetrahydrofuran Complex

Specifications

CAS 462-34-0
Melting Point 11°C to 13°C
Density 1.268 g/mL at 25°C
Boiling Point 180°C
Molecular Formula C4H8BF3O
Linear Formula BF3 -+ THF
MDL Number MFCD00040372
Quantity 100 mL
Molecular Weight (g/mol) 139.91 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.