Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Boron Triiodide

Catalog No. 3076295G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-762-95G 5 g
30-762-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 30-762-95G Supplier Sigma Aldrich Supplier No. 3076295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron triiodide is a strong Lewis acid. It can be prepared by treating iodine (I2) with potassium borohydride (KBH4) in heptane.Boron triiodide can be used as a reagent to cleave C-O bonds in ethers, esters, and alcohols. It can also be used to cleave silanes and halides. Boron triiodide converts alcohols to alkyl iodides; sulfonyl and sulfinyl to disulfides. Borylation of triarylamines can be achievd using this reagent.It can also be used to prepare imidazole based imino aluminum dihalide metal complexes and boron carbonitride (BCN) nanowires.

Specifications

CAS 13517-10-7
Quantity 5 g
Molecular Formula BI3
Linear Formula BI3
MDL Number MFCD00036286
Molecular Weight (g/mol) 391.52 g/mol
Density 3.35 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.