Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boron trioxide | 1303-86-2 | MFCD00011315 | 98.5% | 69.62 g/mol | B2O3 | 500 G
SDP

Catalog No. 5000283672
Encompass_Preferred

Boron trioxide (boric anhydride, B2O3) is a white to off-white solid provided as a high-purity reagent for laboratory and research applications. It is used as a starting material or reagent where low trace-metal content and consistent material properties are required; supporting documentation (data sheet, COA, SDS) is available to confirm specifications.

  • High-purity reagent (purity reported as 98.5% on supplier page).
  • Molecular formula B2O3.
  • Molecular weight 69.62 g/mol.
  • White to off-white solid appearance.
  • Available in 500 G pack size.
  • Data sheet, COA, and SDS available for verification.
  • Intended for research use only.

Catalog No. 50-002-83672 Supplier Medchemexpress LLC Supplier No. HYW129975500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boron trioxide (boric anhydride, B2O3) is a white to off-white solid provided as a high-purity reagent for laboratory and research applications. It is used as a starting material or reagent where low trace-metal content and consistent material properties are required; supporting documentation (data sheet, COA, SDS) is available to confirm specifications.

  • High-purity reagent (purity reported as 98.5% on supplier page).
  • Molecular formula B2O3.
  • Molecular weight 69.62 g/mol.
  • White to off-white solid appearance.
  • Available in 500 G pack size.
  • Data sheet, COA, and SDS available for verification.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.