Learn More
Medchemexpress LLC Boronic acid, B-[1,1'-biphenyl]-2-yl- | 4688-76-0 | 198.03 | 10 G
SDP
Catalog No. 5000447116
Encompass_Preferred
2-Biphenylboronic acid is a biochemical reagent suitable for life science research. It functions as a substrate in palladium-catalyzed cross-coupling reactions and as a reactant in Suzuki-Miyaura cross-coupling reactions. It is also involved in various organic synthesis applications.
  • Substrate for palladium-catalyzed cross-coupling with benzylic acetates
  • Reactant in Suzuki-Miyaura cross-coupling reactions
  • Involved in intramolecular Friedel-Crafts alkylation for chiral tetralins synthesis
  • Used for hydroxylation to phenols
  • Participates in oxidative coupling with alkynes
Catalog No. 50-004-47116 Supplier Medchemexpress LLC Supplier No. HYW00093910G
2-Biphenylboronic acid is a biochemical reagent suitable for life science research. It functions as a substrate in palladium-catalyzed cross-coupling reactions and as a reactant in Suzuki-Miyaura cross-coupling reactions. It is also involved in various organic synthesis applications.
  • Substrate for palladium-catalyzed cross-coupling with benzylic acetates
  • Reactant in Suzuki-Miyaura cross-coupling reactions
  • Involved in intramolecular Friedel-Crafts alkylation for chiral tetralins synthesis
  • Used for hydroxylation to phenols
  • Participates in oxidative coupling with alkynes