Learn More
Medchemexpress LLC Boronic acid, B-[1,1'-biphenyl]-4-yl- | 5122-94-1 | 198.03 | 5 G
SDP
Catalog No. 5000445793
Encompass_Preferred
[1,1'-Biphenyl]-4-ylboronic acid is a biochemical reagent that can be utilized as a biological material or an organic compound for life science-related research. It is a boric acid compound, intended for research use only.
  • Utilized as a biological material
  • Utilized as an organic compound for life science-related research
  • Boric acid compound
  • Tested in HEK293 cells
  • Shows antagonist activity at recombinant human TRPV1 channel (IC50 > 10 μM)
  • Assessed as inhibition of capsaicin-induced Ca2+ flux
Catalog No. 50-004-45793 Supplier Medchemexpress LLC Supplier No. HYW0023645G
[1,1'-Biphenyl]-4-ylboronic acid is a biochemical reagent that can be utilized as a biological material or an organic compound for life science-related research. It is a boric acid compound, intended for research use only.
  • Utilized as a biological material
  • Utilized as an organic compound for life science-related research
  • Boric acid compound
  • Tested in HEK293 cells
  • Shows antagonist activity at recombinant human TRPV1 channel (IC50 > 10 μM)
  • Assessed as inhibition of capsaicin-induced Ca2+ flux