Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boronic acid, B-(3-aminophenyl)- | 30418-59-8 | MFCD00007755 | 99.9% | 136.94 g/mol | C6H8BNO2 | 10 G
SDP

Catalog No. 5000300000
Encompass_Preferred

3-Aminophenylboronic acid is an organoboron reagent and building block used in organic synthesis and biochemical research. It is supplied as a solid of high purity and is commonly used to functionalize nanomaterials, such as copper nanoclusters, enabling rapid, selective detection of glycoproteins.

  • Chemical formula: C6H8BNO2.
  • Molecular weight: 136.94 g/mol.
  • CAS number: 30418-59-8.
  • Purity: 99.87%.
  • Physical form: solid.
  • Applications include biochemical assays and organic synthesis.
  • Available in research pack sizes including 10 g.

Catalog No. 50-003-00000 Supplier Medchemexpress LLC Supplier No. HYW01760410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Aminophenylboronic acid is an organoboron reagent and building block used in organic synthesis and biochemical research. It is supplied as a solid of high purity and is commonly used to functionalize nanomaterials, such as copper nanoclusters, enabling rapid, selective detection of glycoproteins.

  • Chemical formula: C6H8BNO2.
  • Molecular weight: 136.94 g/mol.
  • CAS number: 30418-59-8.
  • Purity: 99.87%.
  • Physical form: solid.
  • Applications include biochemical assays and organic synthesis.
  • Available in research pack sizes including 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.