Learn More
Medchemexpress LLC Boronic acid, B-(3-aminophenyl)- | 30418-59-8 | MFCD00007755 | 99.9% | 136.94 g/mol | C6H8BNO2 | 50 G
SDP
Catalog No. 5000301789
Encompass_Preferred
3-Aminophenylboronic acid (m-aminophenylboronic acid) is an organoboron reagent used in biochemical research and organic synthesis. It functions as a boronate ligand for selective glycoprotein detection and as a versatile building block in cross-coupling and derivatization chemistry. The material is supplied as a high-purity solid suitable for analytical and preparative workflows.
  • High purity suitable for analytical and synthetic applications.
  • Enables boronate-based detection of glycoproteins through selective binding.
  • Serves as a versatile building block for cross-coupling and derivatization reactions.
  • Available in multiple package sizes for small-scale use or scale-up.
  • Provided with datasheet and certificate of analysis for quality assurance.
Catalog No. 50-003-01789 Supplier Medchemexpress LLC Supplier No. HYW01760450G
3-Aminophenylboronic acid (m-aminophenylboronic acid) is an organoboron reagent used in biochemical research and organic synthesis. It functions as a boronate ligand for selective glycoprotein detection and as a versatile building block in cross-coupling and derivatization chemistry. The material is supplied as a high-purity solid suitable for analytical and preparative workflows.
  • High purity suitable for analytical and synthetic applications.
  • Enables boronate-based detection of glycoproteins through selective binding.
  • Serves as a versatile building block for cross-coupling and derivatization reactions.
  • Available in multiple package sizes for small-scale use or scale-up.
  • Provided with datasheet and certificate of analysis for quality assurance.