Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boronic acid, B-(3-cyanophenyl)- | 150255-96-2 | 99.4% | 146.94 | 25 G
SDP

Catalog No. 5000403155
Encompass_Preferred

3-Cyanophenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Purity: 99.42%
  • Molecular weight: 146.94
  • Formula: C7H6BNO2
  • Cas no.: 150255-96-2
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (DMSO): 133.33 mg/mL (907.38 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Catalog No. 50-004-03155 Supplier Medchemexpress LLC Supplier No. 25GHY32927
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Cyanophenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Purity: 99.42%
  • Molecular weight: 146.94
  • Formula: C7H6BNO2
  • Cas no.: 150255-96-2
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (DMSO): 133.33 mg/mL (907.38 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.