Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boronic acid, B-(5-fluoro-2-methoxyphenyl)- | 179897-94-0 | 99.9% | 169.95 | 25 G
SDP

Catalog No. 5000402541
Encompass_Preferred

Boronic acid, B-(5-fluoro-2-methoxyphenyl)- is a solid chemical substance with an appearance ranging from white to off-white. Its 1H NMR spectrum is consistent with its structure, ensuring its identity and quality. This product is suitable for various laboratory applications.

  • Appearance: White to off-white solid
  • 1H NMR spectrum: Consistent with structure
  • Storage stability: Powder stable for 3 years at -20°C and 2 years at 4°C
  • Storage stability: In solvent stable for 6 months at -80°C and 1 month at -20°C

Catalog No. 50-004-02541 Supplier Medchemexpress LLC Supplier No. 25GHY32924
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boronic acid, B-(5-fluoro-2-methoxyphenyl)- is a solid chemical substance with an appearance ranging from white to off-white. Its 1H NMR spectrum is consistent with its structure, ensuring its identity and quality. This product is suitable for various laboratory applications.

  • Appearance: White to off-white solid
  • 1H NMR spectrum: Consistent with structure
  • Storage stability: Powder stable for 3 years at -20°C and 2 years at 4°C
  • Storage stability: In solvent stable for 6 months at -80°C and 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.