Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boronic acid, B-cyclopropyl- | 411235-57-9 | ≥98.0% | C3H7BO2 | 25 G
SDP

Catalog No. 5000359047
Encompass_Preferred

Cyclopropylboronic acid is an intermediate used in the synthesis of EGFR kinase inhibitor (Compound 25). Compound 25 demonstrates antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells, making this product relevant for lung cancer research.

  • Intermediate for synthesizing EGFR kinase inhibitors
  • Supports research on EGFR mutant cells
  • Applicable in lung cancer research
  • Molecular weight: 85.90
  • Appearance: Off-white to yellow solid
  • Soluble in DMSO (≥ 100 mg/mL)

Catalog No. 50-003-59047 Supplier Medchemexpress LLC Supplier No. 25GHY59219
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclopropylboronic acid is an intermediate used in the synthesis of EGFR kinase inhibitor (Compound 25). Compound 25 demonstrates antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells, making this product relevant for lung cancer research.

  • Intermediate for synthesizing EGFR kinase inhibitors
  • Supports research on EGFR mutant cells
  • Applicable in lung cancer research
  • Molecular weight: 85.90
  • Appearance: Off-white to yellow solid
  • Soluble in DMSO (≥ 100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.