Learn More
Medchemexpress LLC Boronic acid, B-(phenyl-2,3,4,5,6-d5) | 215527-70-1 | MFCD02093711 | 100.0% | 126.96 | C6H2D5BO2 | 100 G
SDP
Catalog No. 5000210672
Encompass_Preferred
Phenyl-boronic acid-d5 is the deuterium-labeled analogue of phenylboronic acid used as a research reagent and tracer in chemical and biochemical studies. It is provided as a high-purity isotopologue for analytical and synthetic applications, with specified storage conditions to preserve isotopic integrity.
  • Deuterium-labeled phenylboronic acid (d5) suitable for tracer and analytical studies.
  • High purity by HPLC for reliable analytical results.
  • Defined molecular formula and molecular weight for precise characterization.
  • Recommended storage under nitrogen at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Useful as a reagent in synthesis and isotopic labeling experiments.
Catalog No. 50-002-10672 Supplier Medchemexpress LLC Supplier No. HYW067615100G
Phenyl-boronic acid-d5 is the deuterium-labeled analogue of phenylboronic acid used as a research reagent and tracer in chemical and biochemical studies. It is provided as a high-purity isotopologue for analytical and synthetic applications, with specified storage conditions to preserve isotopic integrity.
  • Deuterium-labeled phenylboronic acid (d5) suitable for tracer and analytical studies.
  • High purity by HPLC for reliable analytical results.
  • Defined molecular formula and molecular weight for precise characterization.
  • Recommended storage under nitrogen at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Useful as a reagent in synthesis and isotopic labeling experiments.