Learn More
Bellco Glass, Inc. Borosil Amber Glass Desiccator with Porcelain Plate and Borosilicate Lid with Plastic Knob, 150 mm D
SDP
Supplier:  Bellco Glass, Inc. 452800746
Encompass_Preferred
Borosil Desiccator Vacuum, Stopcock with PTFE spindle and Porcelain plate, 300 mm, Borosilicate 3.3 glass, CS/1SpecificationsMaterial: GlassGround Flange: 300 mmOverall Dimension: 16.73" L x 16.73" W x 16.93" HBorosilicate Glass 3.3Graduation Range: Type ISteam Sterilization at 121º C is recommended for autoclavingNot recommended for heat applications
Catalog No. 50-255-0016