Learn More
Bellco Glass, Inc. Borosil Beakers, Low-Form, with Spouts, 5,000mL
SDP
Supplier:  Bellco Glass, Inc. 452800919
Encompass_Preferred
Borosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 5000mL CS/4SpecificationsMaterial: GlassBeaker: 5000mLApprox. O.D. x Height: 170mm x 270mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beaker
Catalog No. 50-254-9843