Learn More
Bellco Glass, Inc. Borosil Beakers, Low-Form, with Spouts, 50mL
SDP
Catalog No. 502549854
Encompass_Preferred
Borosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 50mL CS/60SpecificationsMaterial: GlassBeaker: 50mLApprox. O.D. x Height: 42mm x 60mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.
Catalog No. 50-254-9854 Supplier Bellco Glass, Inc. Supplier No. 452800908
Borosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 50mL CS/60SpecificationsMaterial: GlassBeaker: 50mLApprox. O.D. x Height: 42mm x 60mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.