Learn More
Bellco Glass, Inc. Borosil Beakers, Low-Form, with Spouts, 2,000mL
SDP
Supplier:  Bellco Glass, Inc. 452800917
Encompass_Preferred
Borosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 2000mL CS/6SpecificationsMaterial: GlassBeaker: 2000mLApprox. O.D. x Height: 130mm x 185mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.
Catalog No. 50-254-9845