Learn More
Bellco Glass, Inc. Borosil Beakers, Low-Form, with Spouts, 1,000mL
SDP
Supplier:  Bellco Glass, Inc. 452800916
Encompass_Preferred
Griffin Beaker in Low Form with Spout Made of Borosil, 1000mLBorosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 1000mL CS/20SpecificationsMaterial: GlassBeaker: 1000mLApprox. O.D. x Height: 105mm x 145mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.
Catalog No. 50-254-9846