Learn More
Bellco Glass, Inc. Borosil Beakers, Low-Form, with Spouts, 600mL
SDP
Supplier:  Bellco Glass, Inc. 452800914
Encompass_Preferred
Borosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 600mL CS/40SpecificationsMaterial: GlassBeaker: 600mLApprox. O.D. x Height: 90mm x 125mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.
Catalog No. 50-254-9848