Learn More
Bellco Glass, Inc. Borosil Beakers, Low-Form, with Spouts, 500mL
SDP
Catalog No. 502549849
Encompass_Preferred
Borosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 500mL CS/40SpecificationsMaterial: GlassBeaker: 500mLApprox. O.D. x Height: 83mm x 115mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.
Catalog No. 50-254-9849 Supplier Bellco Glass, Inc. Supplier No. 452800913
Borosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 500mL CS/40SpecificationsMaterial: GlassBeaker: 500mLApprox. O.D. x Height: 83mm x 115mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.