Learn More
Bellco Glass, Inc. Borosil Bottles, BOD, 300mL
SDP
Catalog No. 502550073
Encompass_Preferred
Biological Oxygen Demand Bottle (BOD), Borosil , 300mlBOD Bottle with Interchangeable stopper and plastic cap, 300 mLSPECIFICATIONSMaterial: GlassBottle: 300 mLOverall Dimension: 31.89" L x 12.20" W x 6.50" HBorosilicate Glass 3.3Graduation Range: Type IQuantity: Case of 30
Catalog No. 50-255-0073 Supplier Bellco Glass, Inc. Supplier No. 452800689
Biological Oxygen Demand Bottle (BOD), Borosil , 300mlBOD Bottle with Interchangeable stopper and plastic cap, 300 mLSPECIFICATIONSMaterial: GlassBottle: 300 mLOverall Dimension: 31.89" L x 12.20" W x 6.50" HBorosilicate Glass 3.3Graduation Range: Type IQuantity: Case of 30