Learn More
Bellco Glass, Inc. Borosil Bottles, Reagent, Clear, with Stoppers, 500mL, 24/29
SDP
Supplier:  Bellco Glass, Inc. 452800694
Encompass_Preferred
Borosil Reagent Bottles, Plain, Narrow Mouth, Graduated with Interchangeable Flat Head Solid Glass Stopper ISO 4796-2 Ground neck Borosilicate glass Durable Matt finish enamelled area for marking 500 mL CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 500 mLColor: ClearApprox O.D. x Length: 86mm x 180mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A
Catalog No. 50-255-0068