Learn More
Bellco Glass, Inc. Borosil Bottles, Reagent, Clear, with Stoppers, 1,000mL, 29/32
SDP
Catalog No. 502550067
Encompass_Preferred
Borosil Reagent Bottles, Plain, Narrow Mouth, Graduated with Interchangeable Flat Head Solid Glass Stopper ISO 4796-2 Ground neck Borosilicate glass Durable Matt finish enameled area for marking 1L CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 1LColor: ClearDimension: 100mm x 230mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A
Catalog No. 50-255-0067 Supplier Bellco Glass, Inc. Supplier No. 452800695
Borosil Reagent Bottles, Plain, Narrow Mouth, Graduated with Interchangeable Flat Head Solid Glass Stopper ISO 4796-2 Ground neck Borosilicate glass Durable Matt finish enameled area for marking 1L CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 1LColor: ClearDimension: 100mm x 230mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A