Learn More
Bellco Glass, Inc. Borosil Bottles, Weighing, Low Form, 40mL, 60mm x 40mm (OD x H)
SDP
Catalog No. 502550051
Encompass_Preferred
Borosil Weighing Bottles with Interchangeable Stopper, 40 mL, CS/10These bottles are mechanically strong and chemical resistant. Complies with IS 1574.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 40 mLClass/ Quality Grade: Type I, Class ANeck Type: StopperApprox O.D. x Height: 60mm x 40mmQuantity Per Case:10
Catalog No. 50-255-0051 Supplier Bellco Glass, Inc. Supplier No. 452800711
Borosil Weighing Bottles with Interchangeable Stopper, 40 mL, CS/10These bottles are mechanically strong and chemical resistant. Complies with IS 1574.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 40 mLClass/ Quality Grade: Type I, Class ANeck Type: StopperApprox O.D. x Height: 60mm x 40mmQuantity Per Case:10