Learn More
Bellco Glass, Inc. Borosil Bottles, Weighing, Tall Form, 25mL, 30mm x 65mm (OD x H)
SDP
Supplier:  Bellco Glass, Inc. 452800710
Encompass_Preferred
Borosil Weighing Bottles with Interchangeable Stopper 25 mL CS/10These bottles are mechanically strong and chemical resistant. Complies with IS 1574.SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 25 mLClass/ Quality Grade: Type I, Class ANeck Type: StopperApprox O.D. x Height: 30mm x 65mmQuantity Per Case:10
Catalog No. 50-255-0052