Learn More
Bellco Glass, Inc. Borosil Burettes, Class A, Boroflo PTFE Screw Thread, 25mL x 0.10mL, Indiv. Cert
SDP
Supplier:  Bellco Glass, Inc. 452800933
Encompass_Preferred
Borosil Burettes, Boroflo fitted with Boroflo General Purpose Screw Thread PTFE Keys Accuracy as per Class A, ASTM E287-2 with Individual Certificate, 25 mL, CS/ 5Burettes are generally used in the Titration process. Boroflo screw thread stopcocks with PTFE key that is highly resistant to chemical attack. Lubrication is never required, ensuring no seizure or contaminating greases. Excellent flow control with easy cleaning capabilities. Consistent performance between 0 C and 50 C.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 25 mLGraduation Range: 0- 25 mLGraduation Interval: 0.05 mLGraduation Color: Ceramic WhiteClass/ Quality Grade: Type I, Class ATolerance: 0.03 mLQuantity Per Case: 5Accuracy as per ASTM E 287-2 with Individual Batch Certificate
Catalog No. 50-254-9829