Learn More
Bellco Glass, Inc. Borosil Burettes, Class A, Boroflo PTFE Screw Thread, 50mL x 0.10mL, Batch Cert
SDP
Catalog No. 502549828
Encompass_Preferred
Borosil Borosil Burettes, Boroflo fitted with Boroflo General Purpose Screw Thread PTFE Keys Accuracy as per Class A, ASTM E287-2 with Batch Certificate 50 mL Borosilicate 3.3 CS/ 5SpecificationsMaterial: 3.3 Borosilicate Glass Capacity: 50mLColor: ClearOverall Dimension: 32.83" L x 6.50" W x 9.61" HGraduation Color: Ceramic WhiteClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9828 Supplier Bellco Glass, Inc. Supplier No. 452800934
Borosil Borosil Burettes, Boroflo fitted with Boroflo General Purpose Screw Thread PTFE Keys Accuracy as per Class A, ASTM E287-2 with Batch Certificate 50 mL Borosilicate 3.3 CS/ 5SpecificationsMaterial: 3.3 Borosilicate Glass Capacity: 50mLColor: ClearOverall Dimension: 32.83" L x 6.50" W x 9.61" HGraduation Color: Ceramic WhiteClass/Quality Grade: Type 1, Class A