Learn More
Bellco Glass, Inc. Borosil Condensers, Liebig, 200mm Jacket, 19/26
SDP
Supplier:  Bellco Glass, Inc. 452800713
Encompass_Preferred
Borosil Liebig Condenser with Drip Tip, Interchangeable Inner and Outer Joint, 200 MM, CS/5Simple design with a low cooling capacity. The light wall of the inner tube facilitates efficient heat transfer. Specifically used for distillate separation.SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearJacket Length: 200 mmClass/ Quality Grade: Type I, Class ANeck Type: JointsInterchangeable Joints: Socket- 19/26, Cone- 19/26Quantity per Case: 5
Catalog No. 50-255-0049