Learn More
Bellco Glass, Inc. Borosil Cylinders, Class A, TC, 500mL x 5.0mL, Individual Cert
SDP
Supplier:  Bellco Glass, Inc. 452800955
Encompass_Preferred
Borosil, Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Individual Certificate, with Hexagonal Base, Amber Graduations, 500 mL, CS/ 4Borosil Measuring Cylinders are used to receive and measure different amounts of liquids. Built with heavy uniform wall tubing and a strong stable hexagonal base, these cylinders are made to last. Complies with ASTM E 1272 and comes with an individual calibration certificate. Each graduated cylinder is manufactured with chemically resistant 3.3 borosilicate glass with amber enamel graduations. Material: 3.3 Borosilicate GlassColor: ClearCapacity: 500 mLGraduation Range: 5- 500 mLGraduation Interval: 5 mLGraduation Color: AmberClass/ Quality Grade: Type I, Class ATolerance: 2 mLQuantity Per Case: 4Accuracy as per ASTM E 1272 with Individual Certificate
Catalog No. 50-254-9807