Learn More
Bellco Glass, Inc. Borosil Cylinders, Class A, TC, 250mL x 2.0mL, Batch Cert
SDP
Catalog No. 502549810
Encompass_Preferred
Borosil, Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Batch Certificate, with Hexagonal Base, Borosilicate, CS/5, To Contain, Amber Graduation, 250 mLSpecificationsMaterial: GlassSize: 250mLBase: HexagonColor: ClearGraduation Color: AmberOverall Dimension: 22.44" L x 22.44" W x 16.54" HBorosilicate Glass 3.3Graduation Range: Type I, Class A
Catalog No. 50-254-9810 Supplier Bellco Glass, Inc. Supplier No. 452800952
Borosil, Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Batch Certificate, with Hexagonal Base, Borosilicate, CS/5, To Contain, Amber Graduation, 250 mLSpecificationsMaterial: GlassSize: 250mLBase: HexagonColor: ClearGraduation Color: AmberOverall Dimension: 22.44" L x 22.44" W x 16.54" HBorosilicate Glass 3.3Graduation Range: Type I, Class A