Learn More
Bellco Glass, Inc. Borosil Desiccator with cover and Porcelain plate plastic knob 200 mm borosilicate glass
SDP
Supplier:  Bellco Glass, Inc. 452800738
Encompass_Preferred
Borosil Desiccator with cover and Porcelain Plate, Rubber Knob as per IS 6128 Borosilicate 3.3 glass, 150 mmSpecificationsMaterial: 3.3 Borosilicate Glass3082042Overall Dimension: 9.65" L x 9.65" W x 10.83" HClass/Quality Grade: Type 1Steam Sterilization at 121º C is recommended for autoclavingNot recommended for heat applications
Catalog No. 50-255-0024