Learn More
Bellco Glass, Inc. Borosil Desiccators, Standard, 300mm (X-Large)
SDP
Catalog No. 502550022
Encompass_Preferred
Borosil Desiccator with cover and Porcelain Plate, Rubber Knob as per IS 6128 Borosilicate 3.3 glass, 250 mmSpecificationsMaterial: 3.3 Borosilicate Glass Overall Dimension: 14.57" L x 14.57" W x 15.94" H3082044Class/Quality Grade: Type 1Steam Sterilization at 121º C is recommended for autoclavingNot recommended for heat applications
Catalog No. 50-255-0022 Supplier Bellco Glass, Inc. Supplier No. 452800740
Borosil Desiccator with cover and Porcelain Plate, Rubber Knob as per IS 6128 Borosilicate 3.3 glass, 250 mmSpecificationsMaterial: 3.3 Borosilicate Glass Overall Dimension: 14.57" L x 14.57" W x 15.94" H3082044Class/Quality Grade: Type 1Steam Sterilization at 121º C is recommended for autoclavingNot recommended for heat applications