Learn More
Bellco Glass, Inc. Borosil Desiccators, Vacuum, 300mm (X-Large)
SDP
Supplier:  Bellco Glass, Inc. 452800745
Encompass_Preferred
Borosil Desiccator Vacuum, Stopcock with PTFE spindle and Porcelain plate, 250 mm, Borosilicate 3.3 glass, CS/1SpecificationsMaterial: GlassGround Flange: 250 mmOverall Dimension: 14.57" L x 14.57" W x 15.94" HBorosilicate Glass 3.3Graduation Range: Type ISteam Sterilization at 121º C is recommended for autoclavingNot recommended for heat applications
Catalog No. 50-255-0017